missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ferritin Light Chain Monoclonal Antibody (H19), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543827
This item is not returnable.
View return policy
Description
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.
Specifikationer
| Ferritin Light Chain | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02792, P02793 | |
| FTL, Ftl1 | |
| Ser2-Asp175 (Accession P02792), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| H19 | |
| Unconjugated | |
| FTL | |
| ferritin L subunit; Ferritin L subunit 1; ferritin L-chain; ferritin light chain; Ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril; Ftl; Ftl1; Ftl-1; L-ferritin; LFTD; NBIA3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2512, 29292 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering