missing translation for 'onlineSavingsMsg'
Learn More

Ferric Carboxymaltose (Technical Grade), TRC

Product Code. 30138597
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138597 10 g 10g
30138545 1 g 1g
30008122 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138597 Supplier Toronto Research Chemicals Supplier No. TRCF30753010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53858-86-9
Molecular Formula C12H25FeO14
Molecular Weight (g/mol) 449.16
Synonym Ferric Hydroxide Polymaltose Complex, Ferrummate
IUPAC Name iron(3+);(2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;trihydroxide
SMILES [OH-].[OH-].[OH-].[Fe+3].OC[C@H]1O[C@H](OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H22O11.Fe.3H2O/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12;;;;/h1,4-12,14-21H,2-3H2;;3*1H2/q;+3;;;/p-3/t4-,5+,6+,7+,8+,9+,10-,11+,12-;;;;/m0..../s1
Formula Weight 449.0594
Chemical Name or Material Ferric Carboxymaltose (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.