missing translation for 'onlineSavingsMsg'
Learn More

Fenclofenac Glucuronide, TRC

Product Code. 30396132
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396132 1 mg
30406290 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396132 Supplier Toronto Research Chemicals Supplier No. TRCF2475201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77182-37-7
Molecular Formula C20H18Cl2O9
Molecular Weight (g/mol) 473.26
Synonym 1-[2-(2,4-Dichlorophenoxy)benzeneacetate]β-D-Glucopyranuronic Acid, Fenclofenac Ester Glucuronide
SMILES O=C(O[C@@H]1O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]1O)CC2=CC=CC=C2OC3=CC=C(Cl)C=C3Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H18Cl2O9/c21-10-5-6-13(11(22)8-10)29-12-4-2-1-3-9(12)7-14(23)30-20-17(26)15(24)16(25)18(31-20)19(27)28/h1-6,8,15-18,20,24-26H,7H2,(H,27,28)/t15-,16-,17+,18-,20+/m0/s1
Chemical Name or Material Fenclofenac Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.