CAS RN 77182-37-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fenclofenac Glucuronide, TRC
CAS: 77182-37-7 Molecular Formula: C20H18Cl2O9 Molecular Weight (g/mol): 473.26 Synonym: 1-[2-(2,4-Dichlorophenoxy)benzeneacetate]β-D-Glucopyranuronic Acid,Fenclofenac Ester Glucuronide SMILES: O=C(O[C@@H]1O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]1O)CC2=CC=CC=C2OC3=CC=C(Cl)C=C3Cl