missing translation for 'onlineSavingsMsg'
Learn More

Fenchone, TRC

Product Code. 30395229
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395229 100 mg
30404195 1 g
30406986 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395229 Supplier Toronto Research Chemicals Supplier No. TRCF247450100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-79-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.23
Synonym Fenchone, Bicyclo[2.2.1]heptan-2-one, 1,3,3-trimethyl-, 2-Norbornanone, 1,3,3-trimethyl- (6CI,8CI), 1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one, (±)-Fenchone, 1,3,3-Trimethyl-2-norbornanone, 1,3,3-Trimethylnorcamphor, Fenchone, NSC 122687, NSC 8896, dl-Fenchone, Fenchone, Bicyclo[2.2.1]heptan-2-one, 1,3,3-trimethyl-, 2-Norbornanone, 1,3,3-trimethyl- (6CI,8CI), 1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one, (±)-Fenchone, 1,3,3-Trimethyl-2-norbornanone, 1,3,3-Trimethylnorcamphor, NSC 122687, NSC 8896, dl-Fenchone
IUPAC Name 2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC12CCC(C1)C(C)(C)C2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H16O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7H,4-6H2,1-3H3
Formula Weight 152.1201
Chemical Name or Material Fenchone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.