CAS RN 1195-79-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fenchone, TRC
CAS: 1195-79-5 Molecular Formula: C10H16O Molecular Weight (g/mol): 152.23 Synonym: Fenchone,Bicyclo[2.2.1]heptan-2-one, 1,3,3-trimethyl-,2-Norbornanone, 1,3,3-trimethyl- (6CI,8CI),1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one,(±)-Fenchone,1,3,3-Trimethyl-2-norbornanone,1,3,3-Trimethylnorcamphor,Fenchone,NSC 122687,NSC 8896,dl-Fenchone,Fenchone,Bicyclo[2.2.1]heptan-2-one, 1,3,3-trimethyl-,2-Norbornanone, 1,3,3-trimethyl- (6CI,8CI),1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one,(±)-Fenchone,1,3,3-Trimethyl-2-norbornanone,1,3,3-Trimethylnorcamphor,NSC 122687,NSC 8896,dl-Fenchone IUPAC Name: 2,2,4-trimethylbicyclo[2.2.1]heptan-3-one SMILES: CC12CCC(C1)C(C)(C)C2=O