missing translation for 'onlineSavingsMsg'
Learn More

Fenaminstrobin (E/Z Mixture), TRC

Product Code. 30392738
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392738 5 mg 5mg
30402982 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392738 Supplier Toronto Research Chemicals Supplier No. TRCF2456005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353736-75-1
Molecular Formula C21H21Cl2N3O3
Molecular Weight (g/mol) 434.32
Synonym (αe)-2-[[[(E)-[(2E)-3-(2,6-Dichlorophenyl)-1-Methyl-2-Propen-1-Ylidene]Amino]Oxy]Methyl]-α-(Methoxyimino)-N-Methylbenzeneacetamide
SMILES C(C(NC)=O)(=NOC)C1=C(CO/N=C(/C=C/C2=C(Cl)C=CC=C2Cl)\C)C=CC=C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H21Cl2N3O3/c1-14(11-12-17-18(22)9-6-10-19(17)23)25-29-13-15-7-4-5-8-16(15)20(26-28-3)21(27)24-2/h4-12H,13H2,1-3H3,(H,24,27)/b12-11+,25-14-,26-20?
Chemical Name or Material Fenaminstrobin (E/Z Mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.