CAS RN 353736-75-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fenaminstrobin (E/Z Mixture), TRC
CAS: 353736-75-1 Molecular Formula: C21H21Cl2N3O3 Molecular Weight (g/mol): 434.32 Synonym: (αe)-2-[[[(E)-[(2E)-3-(2,6-Dichlorophenyl)-1-Methyl-2-Propen-1-Ylidene]Amino]Oxy]Methyl]-α-(Methoxyimino)-N-Methylbenzeneacetamide SMILES: C(C(NC)=O)(=NOC)C1=C(CO/N=C(/C=C/C2=C(Cl)C=CC=C2Cl)\C)C=CC=C1