missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEN-1 Rabbit anti-Human, Mouse, Clone: 3H9L9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | FEN-1 |
|---|---|
| Clone | 3H9L9 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18334723
|
Novus Biologicals
NBP3-16730-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352192
|
Novus Biologicals
NBP3-16730-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FEN-1 Monoclonal antibody specifically detects FEN-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| FEN-1 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human FEN-1 (P39748). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3H9L9 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2237 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title