missing translation for 'onlineSavingsMsg'
Learn More

FEN-1 Rabbit anti-Human, Mouse, Clone: 3H9L9, Novus Biologicals™

Product Code. 18352192 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352192 100 μg 100µL
18334723 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18352192 Supplier Novus Biologicals Supplier No. NBP316730100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FEN-1 Monoclonal antibody specifically detects FEN-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEN-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3H9L9
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human FEN-1 (P39748).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases
Primary or Secondary Primary
Gene ID (Entrez) 2237
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.