missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline IL-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £343.00
Specifications
| Antigen | IL-6 |
|---|---|
| Clone | 341031 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743828
![]() |
R&D Systems
MAB23051-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18122321
|
R&D Systems
MAB23051 |
100 μg |
£343.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-6 Monoclonal specifically detects IL-6 in Feline samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| IL-6 | |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Feline | |
| P41683 | |
| 3569 | |
| E. coli-derived recombinant feline IL-6 Thr28-Met208 (Glu133Lys) Accession # P41683 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant feline IL-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-6, recombinant mouse (rm) IL-6, recombinant porcine IL-6, recombinant rat (rr) IL-6, recombinant canine IL-6, recombinant cotton rat IL-6, recombinant equine IL-6, rmIL-11, rmLIF, rmOSM, rhCLC or rrCNTF is observed. |
| 341031 | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| IL6 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title