missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline IL-6 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB23051
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Feline samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| IL-6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P41683 | |
| IL6 | |
| E. coli-derived recombinant feline IL-6 Thr28-Met208 (Glu133Lys) Accession # P41683 | |
| 100 μg | |
| Primary | |
| Detects recombinant feline IL-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-6, recombinant mouse (rm) IL-6, recombinant porcine IL-6, recombinant rat (rr) IL-6, recombinant canine IL-6, recombinant cotton rat IL-6, recombinant equine IL-6, rmIL-11, rmLIF, rmOSM, rhCLC or rrCNTF is observed. | |
| Feline | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 341031 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3569 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction