missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FDFT1 Monoclonal Antibody (24GB5645)
GREENER_CHOICE

Mouse Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen FDFT1
Clone 24GB5645
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276289
GREENER_CHOICE
View Documents
Invitrogen™
MA557398
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275294
View Documents
Invitrogen™
MA557399
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FDFT1 Monoclonal Antibody for Western Blot

FDFT1 catalyzes the first step in the cholesterol biosynthetic pathway, the conversion of trans-farnesyldiphosphate to squalene. The loss of promoter activity and response to sterols for FDFT1 is localized to a 69-bp section positioned 131 bp 5-prime to the transcription start site. Sequence analysis of this region shows that it contains a sterol regulatory element-1 (SRE1) previously identified in other sterol regulated genes and 2 putative NF1 binding sites.
Specifications

Specifications

FDFT1
-20°C
Monoclonal
Liquid
IgG2b
Human
FDFT1
DGPT; EGK_18680; ERG9; farnesyl diphosphate farnesyl transferase 1; farnesyl diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase 1; FDFT1; FPP:FPP farnesyltransferase; presqualene-di-diphosphate synthase; SQS; Squalene synthase; squalene synthetase; SS
FDFT1
Primary
Affinity chromatography
24GB5645
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P37268
2222
Recombinant protein of human FDFT1 (aa 1-417).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.