missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FcgR4/CD16-2 Antibody (011), Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £353.00
Specifications
| Antigen | FcgR4/CD16-2 |
|---|---|
| Clone | 011 |
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645565
|
Novus Biologicals
NBP3-05889-100ul |
100 μg |
£353.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696975
|
Novus Biologicals
NBP3-05889-50ul |
50 μg |
£222.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FcgR4/CD16-2 Monoclonal antibody specifically detects FcgR4/CD16-2 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| FcgR4/CD16-2 | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 246256 | |
| IgG | |
| Protein A purified |
| 011 | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| 4833442P21Rik;CD16-2;Fcgam;FcgammaRIV;Fcgr;Fcgr3a;FcgRIV;Fcrl3;Low affinity immunoglobulin gamma Fc region receptor III-A | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FcgR4/CD16-2 (Accession#: NP_653142.2; Met 1-Gln 203). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title