missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FcgR4/CD16-2 Antibody (011), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-05889-100ul
This item is not returnable.
View return policy
Description
FcgR4/CD16-2 Monoclonal antibody specifically detects FcgR4/CD16-2 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| FcgR4/CD16-2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 246256 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 011 | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:100-1:500 | |
| 4833442P21Rik;CD16-2;Fcgam;FcgammaRIV;Fcgr;Fcgr3a;FcgRIV;Fcrl3;Low affinity immunoglobulin gamma Fc region receptor III-A | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FcgR4/CD16-2 (Accession#: NP_653142.2; Met 1-Gln 203). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction