missing translation for 'onlineSavingsMsg'
Learn More

FCGR2A / CD32a Mouse anti-Human, Clone: 1D3F1, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen FCGR2A / CD32a
Clone 1D3F1
Concentration 1 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16808495
missing translation for 'viewDocuments'
Proteintech
66529-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
16898485
missing translation for 'viewDocuments'
Proteintech
66529-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

FCGR2A / CD32a
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CD32, CD32A, CDw32, Fc gamma RII a, Fc gamma RIIa, FCG2, FcGR, FCGR2, FCGR2A, FCGR2A1, FcRII a, IGFR2, IgG Fc receptor II a
FCGR2A
IgG2b
Protein A
Antibody
1D3F1
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
P12318
2212
FCGR2A / CD32a Fusion Protein Ag25878
Primary
-20°C
FCGR2A
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.