missing translation for 'onlineSavingsMsg'
Learn More

FCGR2A / CD32a Mouse anti-Human, Clone: 1D3F1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66529-1-IG-20UL

 View more versions of this product

Product Code. 16808495

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

FCGR2A / CD32a
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P12318
FCGR2A
FCGR2A / CD32a Fusion Protein Ag25878
20 μL
Primary
Human
Antibody
IgG2b
Western Blot, Immunohistochemistry (Paraffin)
1D3F1
Unconjugated
FCGR2A
CD32, CD32A, CDw32, Fc gamma RII a, Fc gamma RIIa, FCG2, FcGR, FCGR2, FCGR2A, FCGR2A1, FcRII a, IGFR2, IgG Fc receptor II a
Mouse
Protein A
RUO
2212
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.