missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc epsilon RI beta/MS4A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | Fc epsilon RI beta/MS4A2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18325955
|
Novus Biologicals
NBP3-02954-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18393367
|
Novus Biologicals
NBP3-02954-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc epsilon RI beta/MS4A2 Polyclonal specifically detects Fc epsilon RI beta/MS4A2 in Human samples. It is validated for Western Blot.Specifications
| Fc epsilon RI beta/MS4A2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| APY, ATOPY, Fc epsilon receptor I beta-chain, FCER1BIGEL, FCERI, High affinity immunoglobulin epsilon receptor beta-subunit (FcERI) (IgE Fcreceptor, beta-subunit) (Fc epsilon receptor I beta-chain), high affinity immunoglobulin epsilon receptor subunit beta, IgE Fc receptor subunit beta, IgE responsiveness (atopic), IGER, IGHER, immunoglobulin E receptor, high affinity, beta polypeptide, Membrane-spanning 4-domains subfamily A member 2, membrane-spanning 4-domains, subfamily A, member 2 (Fc fragment of IgE, highaffinity I, receptor for; beta polypeptide), MS4A1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Fc epsilon RI beta/MS4A2 (NP_000130.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Asthma, Immunology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 2206 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title