missing translation for 'onlineSavingsMsg'
Learn More

Fc epsilon RI beta/MS4A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18393367 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393367 100 μL 100µL
18325955 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18393367 Supplier Novus Biologicals Supplier No. NBP302954100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Fc epsilon RI beta/MS4A2 Polyclonal specifically detects Fc epsilon RI beta/MS4A2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc epsilon RI beta/MS4A2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias APY, ATOPY, Fc epsilon receptor I beta-chain, FCER1BIGEL, FCERI, High affinity immunoglobulin epsilon receptor beta-subunit (FcERI) (IgE Fcreceptor, beta-subunit) (Fc epsilon receptor I beta-chain), high affinity immunoglobulin epsilon receptor subunit beta, IgE Fc receptor subunit beta, IgE responsiveness (atopic), IGER, IGHER, immunoglobulin E receptor, high affinity, beta polypeptide, Membrane-spanning 4-domains subfamily A member 2, membrane-spanning 4-domains, subfamily A, member 2 (Fc fragment of IgE, highaffinity I, receptor for; beta polypeptide), MS4A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Fc epsilon RI beta/MS4A2 (NP_000130.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Asthma, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2206
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.