missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc epsilon RI Antibody (43160) - BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
£146.00 - £493.00
Specifications
| Antigen | Fc epsilon RI |
|---|---|
| Clone | 43160 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18238994
|
Novus Biologicals
NBP1-43279-0.025MG |
0.025 mg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18728543
|
Novus Biologicals
NBP1-43279-0.1mg |
0.1 mg |
£405.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18259492
|
Novus Biologicals
NBP1-43279-0.5MG |
0.5 mg |
£493.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc epsilon RI Monoclonal specifically detects Fc epsilon RI in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Fc epsilon RI | |
| 0.5 mg/mL | |
| Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Mouse | |
| alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
| FCER1A | |
| IgG | |
| Affinity Purified |
| 43160 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Armenian Hamster | |
| Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 2205 | |
| The immunogen for this antibody was FceRI alpha. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title