missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc epsilon RI Antibody (43160) - BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-43279-0.1mg
This item is not returnable.
View return policy
Description
Fc epsilon RI Monoclonal specifically detects Fc epsilon RI in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Fc epsilon RI | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
| Armenian Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| 43160 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| FCER1A | |
| The immunogen for this antibody was FceRI alpha. | |
| 0.1 mg | |
| Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
| 2205 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction