missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £290.00
Specifications
| Antigen | FBXO6 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18356564
|
Novus Biologicals
NBP3-15332-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18321014
|
Novus Biologicals
NBP3-15332-100UL |
100 μg |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FBXO6 Polyclonal antibody specifically detects FBXO6 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| FBXO6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| FBG2F-box/G-domain protein 2, F-box only protein 6, F-box protein 6, F-box protein Fbx6, F-box protein that recognizes sugar chains 2, Fbx6b, FBX6FBS2F-box protein FBG2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-260 of human FBXO6 (NP_060908.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication | |
| PBS, 50% glycerol, pH7.3 | |
| 26270 | |
| IgG | |
| Affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts