missing translation for 'onlineSavingsMsg'
Learn More

FBXO6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18321014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321014 100 μg 100µL
18356564 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18321014 Supplier Novus Biologicals Supplier No. NBP315332100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FBXO6 Polyclonal antibody specifically detects FBXO6 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO6
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias FBG2F-box/G-domain protein 2, F-box only protein 6, F-box protein 6, F-box protein Fbx6, F-box protein that recognizes sugar chains 2, Fbx6b, FBX6FBS2F-box protein FBG2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 70-260 of human FBXO6 (NP_060908.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 26270
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.