missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO33 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | FBXO33 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18386206
|
Novus Biologicals
NBP3-04977-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18303897
|
Novus Biologicals
NBP3-04977-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FBXO33 Polyclonal specifically detects FBXO33 in Rat samples. It is validated for Western Blot.Specifications
| FBXO33 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 254170 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| BMND12, c14_5247, F-box only protein 33, F-box protein 33, Fbx33 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 346-555 of human FBXO33 (NP_976046.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title