missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO33 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04977-100UL
This item is not returnable.
View return policy
Description
FBXO33 Polyclonal specifically detects FBXO33 in Rat samples. It is validated for Western Blot.
Specifications
| FBXO33 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 254170 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| BMND12, c14_5247, F-box only protein 33, F-box protein 33, Fbx33 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 346-555 of human FBXO33 (NP_976046.1). | |
| 100 μL | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction