missing translation for 'onlineSavingsMsg'
Learn More

FBX09 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£167.00 - £302.00

Specifications

Antigen FBX09
Concentration 0.5 mg/mL
Dilution Western Blot 4-6ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence 5ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18126892
View Documents
Novus Biologicals
NBP2-27364SS
0.025 mg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18120663
View Documents
Novus Biologicals
NBP2-27364
0.1 mg
£302.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FBX09 Polyclonal specifically detects FBX09 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

FBX09
Western Blot 4-6ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence 5ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml
Polyclonal
Purified
RUO
Human
Q9UK97
26268
Synthetic peptide made to an N-terminal sequence of the human FBX09 protein (between amino acids 1-100) [UniProt Q9UK97]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Stem Cell Markers
PBS containing 0.05% BSA. with 0.05% Sodium Azide
Cross-immune reaction antigen 1, dJ341E18.2, F-box only protein 9, F-box protein 9, F-box protein Fbx9, FBX9DKFZp434C0118, KIAA0936, NY-REN-57, Renal carcinoma antigen NY-REN-57, VCIA1
FBXO9
IgG
Protein A purified
Reacts with human. Predicted to cross react with primates by homology (100% homologous to most ape species and 95% homologous to most monkey species).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.