missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBX09 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27364
This item is not returnable.
View return policy
Description
FBX09 Polyclonal specifically detects FBX09 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FBX09 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.05% BSA. with 0.05% Sodium Azide | |
| Cross-immune reaction antigen 1, dJ341E18.2, F-box only protein 9, F-box protein 9, F-box protein Fbx9, FBX9DKFZp434C0118, KIAA0936, NY-REN-57, Renal carcinoma antigen NY-REN-57, VCIA1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reacts with human. Predicted to cross react with primates by homology (100% homologous to most ape species and 95% homologous to most monkey species). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 4-6ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence 5ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml | |
| Q9UK97 | |
| FBXO9 | |
| Synthetic peptide made to an N-terminal sequence of the human FBX09 protein (between amino acids 1-100) [UniProt Q9UK97] | |
| 0.1 mg | |
| Stem Cell Markers | |
| 26268 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction