missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBRS Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

£155.00 - £386.00

Specifications

Antigen FBRS
Concentration 0.5 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17703404
View Documents
Invitrogen™
PA5122002
20 μL
£155.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17793394
GREENER_CHOICE
View Documents
Invitrogen™
PA5122001
100 μL
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fibrosin is a lymphokine secreted by activated lymphocytes that induces fibroblast proliferation.
Specifications

Specifications

FBRS
Western Blot
Unconjugated
Rabbit
Human, Mouse
Q8R089, Q9HAH7
14123, 64319
Met1-Asp250 (Accession Q9HAH7), with N-terminal His Tag.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Fbrs
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Fbrs; FBS; FBS1; fbsh; fibrogenic lymphokine; fibrosin; fibrosin 1; fibrosin-1; FLJ11618; probable fibrosin-1; probable fibrosin-1 long transcript protein
Fbrs
IgG
Antigen affinity chromatography, Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.