missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBRS Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122001
This item is not returnable.
View return policy
Description
Fibrosin is a lymphokine secreted by activated lymphocytes that induces fibroblast proliferation.
Specifications
| FBRS | |
| Polyclonal | |
| Unconjugated | |
| Fbrs | |
| Fbrs; FBS; FBS1; fbsh; fibrogenic lymphokine; fibrosin; fibrosin 1; fibrosin-1; FLJ11618; probable fibrosin-1; probable fibrosin-1 long transcript protein | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 14123, 64319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8R089, Q9HAH7 | |
| Fbrs | |
| Met1-Asp250 (Accession Q9HAH7), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction