missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | FBF1 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858673
|
Proteintech
11531-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848673
|
Proteintech
11531-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is induced by N-terminal (20-347aa) of FBF1. It recognizes all the six isoforms.
Keratin-binding protein required for epithelial cell polarization. Involved in apical junction complex (AJC) assembly via its interaction with PARD3. Required for ciliogenesis.Specifications
| FBF1 | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| A2A870, Q8TES7 | |
| 217335, 85302 | |
| FBF1 Fusion Protein Ag2106 | |
| Primary | |
| -20°C | |
| FBF1 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Alb, albatross, Fas (TNFRSF6) binding factor 1, FBF 1, FBF1, FBF1 Ens, HCG1989313, isoform CRA_b, KIAA1863 | |
| FBF1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel