missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11531-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is induced by N-terminal (20-347aa) of FBF1. It recognizes all the six isoforms.
Keratin-binding protein required for epithelial cell polarization. Involved in apical junction complex (AJC) assembly via its interaction with PARD3. Required for ciliogenesis.Specifications
| FBF1 | |
| Polyclonal | |
| Unconjugated | |
| FBF1 | |
| Alb, albatross, Fas (TNFRSF6) binding factor 1, FBF 1, FBF1, FBF1 Ens, HCG1989313, isoform CRA_b, KIAA1863 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217335, 85302 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A2A870, Q8TES7 | |
| FBF1 | |
| FBF1 Fusion Protein Ag2106 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction