missing translation for 'onlineSavingsMsg'
Learn More

FATP4/SLC27A4 Antibody (11A1), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen FATP4/SLC27A4
Clone 11A1
Dilution Western Blot 1:500-1:2000, ELISA
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30037258
View Documents
Novus Biologicals
NBP3-26320-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30028366
View Documents
Novus Biologicals
NBP3-26320-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FATP4/SLC27A4 Monoclonal antibody specifically detects FATP4/SLC27A4 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications

Specifications

FATP4/SLC27A4
Western Blot 1:500-1:2000, ELISA
Monoclonal
Purified
RUO
PBS, pH 7.4, 150mM NaCl and 50% glycerol
10999
IgG
Affinity purified
11A1
Western Blot, ELISA
Unconjugated
Rabbit
Human, Mouse
ACSVL4Solute carrier family 27 member 4, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-4, FATP4Fatty acid transport protein 4, IPS, long-chain fatty acid transport protein 4, solute carrier family 27 (fatty acid transporter), member 4
A synthesized peptide derived from Human FATP4/SLC27A4 [UniProt Q6P1M0]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.