missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FATP4/SLC27A4 Antibody (11A1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26320-100ul
This item is not returnable.
View return policy
Description
FATP4/SLC27A4 Monoclonal antibody specifically detects FATP4/SLC27A4 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| FATP4/SLC27A4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10999 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 11A1 | |
| Western Blot 1:500-1:2000, ELISA | |
| ACSVL4Solute carrier family 27 member 4, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-4, FATP4Fatty acid transport protein 4, IPS, long-chain fatty acid transport protein 4, solute carrier family 27 (fatty acid transporter), member 4 | |
| A synthesized peptide derived from Human FATP4/SLC27A4 [UniProt Q6P1M0] | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction