Learn More
Fasudil monohydrochloride, 99+%, Thermo Scientific™
£281.00 - £822.00
Chemical Identifiers
| CAS | 105628-07-7 |
|---|---|
| Molecular Formula | C14H18ClN3O2S |
| Molecular Weight (g/mol) | 327.827 |
| MDL Number | MFCD00943198 |
| InChI Key | LFVPBERIVUNMGV-UHFFFAOYSA-N |
| Synonym | fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br |
| PubChem CID | 163751 |
| IUPAC Name | 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride |
| SMILES | C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15443469
|
Thermo Scientific Alfa Aesar
J60751.MD |
250mg |
£281.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15453469
|
Thermo Scientific Alfa Aesar
J60751.ME |
500mg |
£494.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15433469
|
Thermo Scientific Alfa Aesar
J60751.03 |
1g |
£822.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water and dimethyl sulfoxide. Insoluble in ethanol.
Notes
Incompatible with acids, bases and oxidizing agents.
Chemical Identifiers
| 105628-07-7 | |
| 327.827 | |
| LFVPBERIVUNMGV-UHFFFAOYSA-N | |
| 163751 | |
| C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl |
| C14H18ClN3O2S | |
| MFCD00943198 | |
| fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br | |
| 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride |
Specifications
| 105628-07-7 | |
| MFCD00943198 | |
| fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br | |
| LFVPBERIVUNMGV-UHFFFAOYSA-N | |
| 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride | |
| 163751 | |
| ≥99% |
| C14H18ClN3O2S | |
| 14,3942 | |
| Soluble in water and dimethyl sulfoxide. Insoluble in ethanol. | |
| C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl | |
| 327.827 | |
| 327.83 | |
| Fasudil monohydrochloride |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'rtecsNumber' : HM4031166
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.