Learn More
Fasudil monohydrochloride, 99+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J60751.03
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water and dimethyl sulfoxide. Insoluble in ethanol.
Notes
Incompatible with acids, bases and oxidizing agents.
Chemical Identifiers
| 105628-07-7 | |
| 327.827 | |
| LFVPBERIVUNMGV-UHFFFAOYSA-N | |
| 163751 | |
| C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl |
| C14H18ClN3O2S | |
| MFCD00943198 | |
| fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br | |
| 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride |
Specifications
| 105628-07-7 | |
| MFCD00943198 | |
| 14,3942 | |
| Soluble in water and dimethyl sulfoxide. Insoluble in ethanol. | |
| C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl | |
| 327.827 | |
| 327.83 | |
| Fasudil monohydrochloride |
| C14H18ClN3O2S | |
| 1g | |
| fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br | |
| LFVPBERIVUNMGV-UHFFFAOYSA-N | |
| 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride | |
| 163751 | |
| ≥99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.