missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm Fast Firing Furnace

Ideal for simulation of typical fast-firing processes up to a maximum firing temperature of 1300 °C. The combination of high performance, low thermal mass and powerful cooling fans provides for cycle times from cold to cold up to 35 minutes with an opening temperature of approx. 300 °C.

Brand:  Nabertherm LS1213PN1

Additional Details : Weight : 130.00000kg

 View more versions of this product

Product Code. 15958270

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Maximum Temperature: 1300 °C
  • Very compact design
  • Ceramic grid tubes as charge support
  • Floor and lid heating
  • Two-zone control, bottom and lid
  • Integrated cooling fans, programmable to speed up charge cooling including housing cooling
  • Programmable lid opening of approximately 60 mm for faster cooling without activating the fan
  • Thermocouple type S for top and bottom zone
  • Castors for easy furnace moving
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications

Specifications

12 L
350 x 350 x 40 mm
150 kg
1300°C
880 x 750 x 1090 mm
3-phase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers