missing translation for 'onlineSavingsMsg'
Learn More

Farnesal (Mixture of Isomers, Technical Grade), TRC

Product Code. 30399280
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399280 10 mg
30408262 50 mg
30395067 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399280 Supplier Toronto Research Chemicals Supplier No. TRCF10241510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19317-11-4
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.35
Synonym 3,7,11-Trimethyl-2,6,10-dodecatrienal
SMILES CC(=CCC\C(=C\CC\C(=C\C=O)\C)\C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H24O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11-12H,5-6,8,10H2,1-4H3/b14-9+,15-11+
Chemical Name or Material Farnesal (Mixture of Isomers, Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.