CAS RN 19317-11-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Farnesal (Mixture of Isomers, Technical Grade), TRC
CAS: 19317-11-4 Molecular Formula: C15H24O Molecular Weight (g/mol): 220.35 Synonym: 3,7,11-Trimethyl-2,6,10-dodecatrienal SMILES: CC(=CCC\C(=C\CC\C(=C\C=O)\C)\C)C