missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCM Antibody (CV5.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£176.00 - £341.00
Specifications
| Antigen | FANCM |
|---|---|
| Clone | CV5.1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation 2 - 4 μg/mL lysate, Immunohistochemistry-Paraffin, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658228
|
Novus Biologicals
NBP2-50418-0.025ml |
0.025 mL |
£176.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675316
|
R&D Systems
NBP2-50418 |
0.1 mL |
£341.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCM Monoclonal antibody specifically detects FANCM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDownSpecifications
| FANCM | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| DNA Repair, Epigenetics | |
| PBS | |
| 57697 | |
| IgG1 κ | |
| Protein A purified |
| CV5.1 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation 2 - 4 μg/mL lysate, Immunohistochemistry-Paraffin, Knockdown Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATP-dependent RNA helicase FANCM, EC 3.6.1, FAAP250EC 3.6.4.13, Fanconi anemia, complementation group M, Fanconi anemia-associated polypeptide of 250 kDa, KIAA1596Fanconi anemia group M protein, MGC176453, Protein FACM, Protein Hef ortholog | |
| His-tagged denatured FANCM (aa 1507-1679) made in E.coli | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title