missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCM Antibody (CV5.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems NBP2-50418
This item is not returnable.
View return policy
Description
FANCM Monoclonal antibody specifically detects FANCM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown
Specifications
| FANCM | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation 2 - 4 μg/mL lysate, Immunohistochemistry-Paraffin, Knockdown Validated | |
| ATP-dependent RNA helicase FANCM, EC 3.6.1, FAAP250EC 3.6.4.13, Fanconi anemia, complementation group M, Fanconi anemia-associated polypeptide of 250 kDa, KIAA1596Fanconi anemia group M protein, MGC176453, Protein FACM, Protein Hef ortholog | |
| His-tagged denatured FANCM (aa 1507-1679) made in E.coli | |
| 0.1 mL | |
| DNA Repair, Epigenetics | |
| 57697 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| CV5.1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction