missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FANCA Recombinant Rabbit Monoclonal Antibody (23GB5360), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552844
This item is not returnable.
View return policy
Description
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.
Specifications
| FANCA | |
| Recombinant Monoclonal | |
| Unconjugated | |
| FANCA | |
| AW208693; FA; FA1; FAA; FACA; FAH; FA-H; Fanca; FANCH; Fanconi anemia complementation group A; Fanconi anemia group A protein; Fanconi anemia group A protein homolog; Fanconi anemia, complementation group A; Fanconi anemia, complementation group H; Fanconi anemia, type 1; Protein FACA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2175 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB5360 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15360 | |
| FANCA | |
| A synthesized peptide derived from human FANCA (1-130AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur