missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FANCA Recombinant Rabbit Monoclonal Antibody (23GB5360), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | FANCA |
|---|---|
| Clone | 23GB5360 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089723
![]() |
Invitrogen™
MA552844 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087433
|
Invitrogen™
MA552845 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCA Recombinant Monoclonal antibody specifically detects FANCA in Human samples. It is validated for Western BlotSpecifications
| FANCA | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O15360 | |
| 2175 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5360 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AW208693; FA; FA1; FAA; FACA; FAH; FA-H; Fanca; FANCH; Fanconi anemia complementation group A; Fanconi anemia group A protein; Fanconi anemia group A protein homolog; Fanconi anemia, complementation group A; Fanconi anemia, complementation group H; Fanconi anemia, type 1; Protein FACA | |
| A synthesized peptide derived from human FANCA (1-130AA). | |
| Primary | |
| -20°C | |
| FANCA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title