missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ IsoTherm-System™

Product Code. p-8004111 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
0.5mL tubes
1.5/2.0mL tubes
Type:
IsoPack and IsoRack set
IsoSafe and IsoPack
Temperature Range:
-21°C
0°C
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Temperature Range unitSize
10343771 0.5mL tubes IsoPack and IsoRack set 0°C Each
10202271 1.5/2.0mL tubes IsoSafe and IsoPack 0°C Each
10579291 1.5/2.0mL tubes IsoPack and IsoRack set 0°C Each
10459072 0.5mL tubes IsoPack and IsoRack set -21°C Each
10798551 0.5mL tubes IsoSafe and IsoPack -21°C Each
10613751 0.5mL tubes IsoSafe and IsoPack 0°C Each
10604311 1.5/2.0mL tubes IsoPack and IsoRack set -21°C Each
10018550 1.5/2.0mL tubes IsoSafe and IsoPack -21°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10343771 Supplier Eppendorf™ Supplier No. 3880000160

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as an enzyme bar to prevent enzymes from becoming inactivated, as well as for PCR* sample preparation, sample transport and slow thawing of frozen samples.

  • Holds 24 tubes (0.5mL/1.5mL/2.0mL Safe-Lock microcentrifuge tubes or 1.7mL Safe-Twist screw cap tubes)
  • IsoPack™ (cold pack) white for 0°C and blue for -21°C, cooled to refrigeration temperature in freezer compartment
  • Isothermal times of >6hr (0°C, white) and >3hr (-21°C, blue)
  • No more melting ice baths (danger of contamination)
  • All system components can be used separately
  • Racks are stackable, autoclavable, markable, floatable and can be centrifuged in a microtitre-plate rotor Specialises in cooling samples effectively and constantly at -21°C or 0°C over many hours.
TRUSTED_SUSTAINABILITY

Specifications

Type IsoPack and IsoRack set
Capacity (Metric) 0.5 mL
Temperature Range 0°C
For Use With (Equipment) 0.5mL tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.