missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM54A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | FAM54A |
|---|---|
| Concentration | 650 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069845
|
Proteintech
26569-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18068745
|
Proteintech
26569-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FAM54A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FAM54A. This antibody recognizes human antigen. The FAM54A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FAM54A | |
| Western Blot, ELISA | |
| Rabbit | |
| 113115 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 650 μg/mL | |
| Liquid | |
| Human | |
| DUFD1, FAM54A, Mitochondrial fission regulator 2, MTFR2, Protein FAM54A | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title