Learn More
FAM54A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26569-1-AP-150UL
This item is not returnable.
View return policy
Description
The FAM54A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FAM54A. This antibody recognizes human antigen. The FAM54A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FAM54A | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| DUFD1, FAM54A, Mitochondrial fission regulator 2, MTFR2, Protein FAM54A | |
| 150 μL | |
| 113115 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |