missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM50A Recombinant Rabbit Monoclonal Antibody (23GB5830), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen FAM50A
Clone 23GB5830
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089203
View Documents
Invitrogen™
MA552924
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30087881
View Documents
Invitrogen™
MA552925
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FAM50A Recombinant Monoclonal antibody specifically detects FAM50A in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

FAM50A
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
Q14320, Q9WV03
108160, 293862, 9130
IgG
Affinity chromatography
Antibody
23GB5830
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
9F; D0HXS9928E; DXS9928E; FAM50A; family with sequence similarity 50 member A; family with sequence similarity 50, member A; HXC26; HXC-26; Protein FAM50A; Protein HXC-26; Protein XAP-5; RGD1560964; XAP5; XAP-5; XX-FW81657B9.2
A synthesized peptide derived from human FAM50A (1-45AA).
Primary
-20°C
FAM50A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.