missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM50A Recombinant Rabbit Monoclonal Antibody (23GB5830), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552925
This item is not returnable.
View return policy
Description
May be a DNA-binding protein or transcriptional factor.
Specifications
| FAM50A | |
| Recombinant Monoclonal | |
| Unconjugated | |
| FAM50A | |
| 9F; D0HXS9928E; DXS9928E; FAM50A; family with sequence similarity 50 member A; family with sequence similarity 50, member A; HXC26; HXC-26; Protein FAM50A; Protein HXC-26; Protein XAP-5; RGD1560964; XAP5; XAP-5; XX-FW81657B9.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108160, 293862, 9130 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5830 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14320, Q9WV03 | |
| FAM50A | |
| A synthesized peptide derived from human FAM50A (1-45AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction