missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM46C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£147.00 - £216.00
Specifications
| Antigen | FAM46C |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
FAM46C Polyclonal antibody specifically detects FAM46C in Human samples. It is validated for Western BlotSpecifications
| FAM46C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS, pH7.0 | |
| 54855 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| family with sequence similarity 46, member C, FLJ20202, hypothetical protein LOC54855 | |
| Produced in rabbits immunized with E. coli-derived Human FAM46C fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title