missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM46C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12613-100ul
This item is not returnable.
View return policy
Descripción
FAM46C Polyclonal antibody specifically detects FAM46C in Human samples. It is validated for Western Blot
Especificaciones
| FAM46C | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| family with sequence similarity 46, member C, FLJ20202, hypothetical protein LOC54855 | |
| Produced in rabbits immunized with E. coli-derived Human FAM46C fragment | |
| 100 μg | |
| Cell Biology | |
| 54855 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering