missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM46A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | FAM46A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FAM46A Polyclonal specifically detects FAM46A in Human samples. It is validated for Western Blot.Specifications
| FAM46A | |
| Polyclonal | |
| Rabbit | |
| Q96IP4 | |
| 55603 | |
| IgG | |
| 50 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C6orf37, chromosome 6 open reading frame 37, family with sequence similarity 46, member A, FLJ20037, FLJ31495, HBV XAg-transactivated protein 11, HBV X-transactivated gene 11 protein, hypothetical protein LOC55603, retinal expressed gene C6orf37, XTP11 | |
| Synthetic peptides corresponding to FAM46A(family with sequence similarity 46, member A) The peptide sequence was selected from the N terminal of FAM46A. Peptide sequence MAEGEGYFAMSEDELACSPYIPLGGDFGGGDFGGGDFGGGDFGGGGSFGG. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title